missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Phenylhexanoic acid, 98+%

Brand:  Alfa Aesar™ A11187.14

Code : NEW

Additional Details : CAS Number : 5581-75-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11463484

  • £402.50 / 25g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Description

Description

Applications
6-Phenylhexanoic acid was employed as a model compound to investigate the effects of chromophore orientation and molecular conformation on surface-enhanced Raman scattering based on metal nanostructures. It was also employed as a substrate to study the stoichiometry of side chain metabolism or complete mineralization of surrogate napthenic acids under methanogenic conditions

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

6-Phenylhexanoic acid
C6H5(CH2)5CO2H
MFCD00014381
5581-75-9
192.26
1950106

Specifications

6-Phenylhexanoic acid
157° to 160°C (0.3mmHg)
25g
≥98%
192.26
1950106
>110°C (230°F)
17° to 19°C
5581-75-9
C6H5(CH2)5CO2H
MFCD00014381
Videos
SDS