missing translation for 'onlineSavingsMsg'
Learn More

(6-Phenyl-3-pyridinyl)methylamine, 95%, Maybridge

Brand:  Maybridge CC21113DA

Code : 31

Additional Details : CAS Number : 294648-38-7 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10336784

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

(6-Phenyl-3-pyridinyl)methylamine
94.5
95%
C12H12N2
MFCD09025823
294648-38-7
100.0
95%
184.24

Specifications

(6-Phenyl-3-pyridinyl)methylamine
294648-38-7
Amber Glass Bottle
95%
C12H12N2
MFCD09025823
1g
94.5
100.0
95%
184.24
Videos
SDS