missing translation for 'onlineSavingsMsg'
Learn More

6-Phenoxy-3-pyridinamine, ≥95%, Thermo Scientific™

Catalog Number p-2703455
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
25194-67-6
C11H10N2O
186.214
MFCD01692449
DETKIRMPBJPJRQ-UHFFFAOYSA-N
6-phenoxy-3-pyridinamine, 3-amino-6-phenoxypyridine, 5-amino-2-phenoxypyridine, pyridine, 5-amino-2-phenoxy, 6-phenoxypyridine-3-amine, 3-pyridinamine,6-phenoxy, 3-pyridinamine, 6-phenoxy, 6-phenoxy-pyridin-3-ylamine, 6-phenoxy-3-pyridylamine, 2-phenoxy-5-aminopyridine
212856
6-phenoxypyridin-3-amine
C1=CC=C(C=C1)OC2=NC=C(C=C2)N
This item is not returnable. View return policy
25194-67-6
C11H10N2O
186.214
MFCD01692449
DETKIRMPBJPJRQ-UHFFFAOYSA-N
6-phenoxy-3-pyridinamine, 3-amino-6-phenoxypyridine, 5-amino-2-phenoxypyridine, pyridine, 5-amino-2-phenoxy, 6-phenoxypyridine-3-amine, 3-pyridinamine,6-phenoxy, 3-pyridinamine, 6-phenoxy, 6-phenoxy-pyridin-3-ylamine, 6-phenoxy-3-pyridylamine, 2-phenoxy-5-aminopyridine
212856
6-phenoxypyridin-3-amine
C1=CC=C(C=C1)OC2=NC=C(C=C2)N
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 186.21
Percent Purity 95%
Chemical Name or Material 6-Phenoxy-3-pyridinamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'