missing translation for 'onlineSavingsMsg'
Learn More

6-Nitropiperonal, 98+%, ACROS Organics™

Brand:  Acros Organics 160070050

Code : 29

Additional Details : CAS Number : 712-97-0 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 10331344

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

6-Nitropiperonal
712-97-0
100.0
98+%
195.13
98+%
98.0
≥98%
C8H5NO5
MFCD00005829

Specifications

6-Nitropiperonal
712-97-0
100.0
Glass bottle
≥98%
C8H5NO5
195.13
92.0° to 98.0°C
98+%
98.0
Authentic
Fine Crystalline Powder
98+%
Yellow
MFCD00005829
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Nitropiperonal

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS