missing translation for 'onlineSavingsMsg'
Learn More

6-[N-(2-chloro-4-pyrimidinyl)methylamino]-2,3-dimethyl-2H-indazole, 96%, Thermo Scientific™

Catalog Number 15458545
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 444731-75-3
Melting Point 167°C to 173°C
Molecular Formula C14H14ClN5
MDL Number MFCD12923006
Quantity 500 mg
Synonym n-2-chloropyrimidin-4-yl-n,2,3-trimethyl-2h-indazol-6-amine, 2h-indazol-6-amine, n-2-chloro-4-pyrimidinyl-n,2,3-trimethyl, n-2-chloropyrimidin-4-yl-n-methyl-2,3-dimethyl-2h-indazol-6-amine, n-2-chloro-4-pyrimidinyl-n,2,3-trimethyl-2h-indazol-6-amine, 2h-indazol-6-amine,n-2-chloro-4-pyrimidinyl-n,2,3-trimethyl, n-2-chloropyrimidin-4-yl-n,2,3-trimethylindazol-6-amine, 6-n-2-chloro-4-pyrimidinyl methylamino-2,3-dimethyl-2h-indazole, 2-chloro-pyrimidin-4-yl-2,3-dimethyl-2h-indazol-6-yl-methyl-amine, n-2-chloropyrimidin-4-yl-n-m ethyl-2,3-dimethyl-2h-indazol-6-amine
InChI Key DVGMRZQSSNNTFY-UHFFFAOYSA-N
SMILES CC1=C2C=CC(=CC2=NN1C)N(C)C3=NC(=NC=C3)Cl
IUPAC Name N-(2-chloropyrimidin-4-yl)-N,2,3-trimethylindazol-6-amine
Molecular Weight (g/mol) 287.751
PubChem CID 11608962
Formula Weight 287.75
Percent Purity 96%
Chemical Name or Material 6-[N-(2-chloro-4-pyrimidinyl)methylamino]-2,3-dimethyl-2H-indazole
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'