missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Morpholinopyridine-2-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 6-morpholinopicolinaldehyde, 6-morpholinopyridine-2-carbaldehyde, 6-morpholin-4-yl pyridine-2-carbaldehyde, 6-morpholin-4-yl pyridine-2-carboxaldehyde, 4-6-formylpyridin-2-yl morpholine, 2-pyridinecarboxaldehyde,6-4-morpholinyl, 2-pyridinecarboxaldehyde, 6-4-morpholinyl, 6-morpholin-4-yl-pyridine-2-carbaldehyde, 6-morpholin-4-yl-pyridine-2-carboxaldehyde, 6-morpholin-4-yl pyridine-2-carboxaldehyd |
Formula Weight | 192.22 |
Percent Purity | 97% |
Chemical Name or Material | 6-Morpholinopyridine-2-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'