missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 245.67 |
Percent Purity | 97% |
Chemical Name or Material | 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'