missing translation for 'onlineSavingsMsg'
Learn More

6-Methylquinoline, 98%, Thermo Scientific Chemicals

Catalog Number 15254218
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
91-62-3
C10H9N
143.19
MFCD00006804
LUYISICIYVKBTA-UHFFFAOYSA-N
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
7059
6-methylquinoline
CC1=CC=C2N=CC=CC2=C1
This item is not returnable. View return policy
91-62-3
C10H9N
143.19
MFCD00006804
LUYISICIYVKBTA-UHFFFAOYSA-N
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
7059
6-methylquinoline
CC1=CC=C2N=CC=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Orange
Density 1.0600g/mL
Boiling Point 256.0°C to 260.0°C
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6125 to 1.6145
Quantity 100 g
Specific Gravity 1.06
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol
Formula Weight 143.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 6-Methylquinoline, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methylquinoline, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'