missing translation for 'onlineSavingsMsg'
Learn More

(6-Methylpyridin-2-yl)methanol, 97%, Thermo Scientific™

Catalog Number p-2693800
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1122-71-0
C7H9NO
123.16
MFCD00023520
JLVBSBMJQUMAMW-UHFFFAOYSA-N
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
70736
(6-methylpyridin-2-yl)methanol
CC1=CC=CC(CO)=N1
This item is not returnable. View return policy
1122-71-0
C7H9NO
123.16
MFCD00023520
JLVBSBMJQUMAMW-UHFFFAOYSA-N
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
70736
(6-methylpyridin-2-yl)methanol
CC1=CC=CC(CO)=N1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 123.15
Percent Purity 97%
Chemical Name or Material (6-Methylpyridin-2-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'