missing translation for 'onlineSavingsMsg'
Learn More

6-Methylcoumarin, 99%, Thermo Scientific Chemicals

Catalog Number p-7024540
Click to view available options
Quantity:
50 g
100 g
250 g
This item is not returnable. View return policy
92-48-8
C10H8O2
160.17
MFCD00006875
FXFYOPQLGGEACP-UHFFFAOYSA-N
6-methylcoumarin, 6-methylbenzopyrone, 6-methyl-2h-chromen-2-one, toncarine, 6-methyl coumarin, methyl coumarin, 6-methylcoumarinic anhydride, coumarin, 6-methyl, 6-methyl-1,2-benzopyrone, 6-methylcumarin
7092
CHEBI:563586
6-methylchromen-2-one
CC1=CC=C2OC(=O)C=CC2=C1
This item is not returnable. View return policy
92-48-8
C10H8O2
160.17
MFCD00006875
FXFYOPQLGGEACP-UHFFFAOYSA-N
6-methylcoumarin, 6-methylbenzopyrone, 6-methyl-2h-chromen-2-one, toncarine, 6-methyl coumarin, methyl coumarin, 6-methylcoumarinic anhydride, coumarin, 6-methyl, 6-methyl-1,2-benzopyrone, 6-methylcumarin
7092
CHEBI:563586
6-methylchromen-2-one
CC1=CC=C2OC(=O)C=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 74°C to 77°C
Boiling Point 303°C (725 mmHg)
Quantity 50 g
Beilstein 4222
Formula Weight 160.17
Percent Purity 99%
Chemical Name or Material 6-Methylcoumarin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'