missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methyl-2-pyridinecarboxaldehyde, 97%, ACROS Organics™
£27.53 - £295.00
Chemical Identifiers
CAS | 1122-72-1 |
---|---|
Molecular Formula | C7H7NO |
Molecular Weight (g/mol) | 121.14 |
MDL Number | MFCD00006291 |
InChI Key | AHISYUZBWDSPQL-UHFFFAOYSA-N |
Synonym | 6-methyl-2-pyridinecarboxaldehyde, 6-methylpicolinaldehyde, 2-formyl-6-methylpyridine, 6-methylpyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 6-methyl, 6-methylpyridine-2-aldehyde, 2-formyl-6-picoline, 6-methyl-2-pyridinecarbaldehyde, 6-methyl-2-picolinaldehyde, 6-methylpyridyl-2-aldehyde |
PubChem CID | 70737 |
IUPAC Name | 6-methylpyridine-2-carbaldehyde |
SMILES | CC1=CC=CC(=N1)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10791725
|
Acros Organics
127570010 |
1g | Glass bottle |
£27.53
1g |
In stock, ready to ship: 2 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10429481
|
Acros Organics
127570050 |
5g | Glass bottle |
£89.21
5g |
Estimated Shipment 09-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10035953
|
Acros Organics
127570250 |
25g | Glass bottle |
£295.00
25g |
Estimated Shipment 09-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
1122-72-1 | |
121.14 | |
AHISYUZBWDSPQL-UHFFFAOYSA-N | |
70737 | |
CC1=CC=CC(=N1)C=O |
C7H7NO | |
MFCD00006291 | |
6-methyl-2-pyridinecarboxaldehyde, 6-methylpicolinaldehyde, 2-formyl-6-methylpyridine, 6-methylpyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 6-methyl, 6-methylpyridine-2-aldehyde, 2-formyl-6-picoline, 6-methyl-2-pyridinecarbaldehyde, 6-methyl-2-picolinaldehyde, 6-methylpyridyl-2-aldehyde | |
6-methylpyridine-2-carbaldehyde |
Specifications
6-Methyl-2-pyridinecarboxaldehyde | |
Authentic | |
1.527 | |
31°C to 33°C | |
97% | |
96.0 | |
96% min. (GC) | |
CH(C5H3N)COH | |
6-methyl-2-pyridinecarboxaldehyde, 6-methylpicolinaldehyde, 2-formyl-6-methylpyridine, 6-methylpyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 6-methyl, 6-methylpyridine-2-aldehyde, 2-formyl-6-picoline, 6-methyl-2-pyridinecarbaldehyde, 6-methyl-2-picolinaldehyde, 6-methylpyridyl-2-aldehyde | |
AHISYUZBWDSPQL-UHFFFAOYSA-N | |
6-methylpyridine-2-carbaldehyde | |
70737 | |
Low Melting Solid |
68°C | |
Glass bottle | |
Yellow to Brown | |
1g | |
1122-72-1 | |
100.0 | |
C7H7NO | |
MFCD00006291 | |
Solubility in water: 750g/L water (20°C). Other solubilities: soluble in ether and acetone | |
CC1=CC=CC(=N1)C=O | |
121.14 | |
121.14 | |
97% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 6-Methyl-2-pyridinecarboxaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.