missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-pyridinecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 10791725
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1122-72-1
C7H7NO
121.14
MFCD00006291
AHISYUZBWDSPQL-UHFFFAOYSA-N
6-methyl-2-pyridinecarboxaldehyde, 6-methylpicolinaldehyde, 2-formyl-6-methylpyridine, 6-methylpyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 6-methyl, 6-methylpyridine-2-aldehyde, 2-formyl-6-picoline, 6-methyl-2-pyridinecarbaldehyde, 6-methyl-2-picolinaldehyde, 6-methylpyridyl-2-aldehyde
70737
6-methylpyridine-2-carbaldehyde
CC1=CC=CC(=N1)C=O
This item is not returnable. View return policy
1122-72-1
C7H7NO
121.14
MFCD00006291
AHISYUZBWDSPQL-UHFFFAOYSA-N
6-methyl-2-pyridinecarboxaldehyde, 6-methylpicolinaldehyde, 2-formyl-6-methylpyridine, 6-methylpyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 6-methyl, 6-methylpyridine-2-aldehyde, 2-formyl-6-picoline, 6-methyl-2-pyridinecarbaldehyde, 6-methyl-2-picolinaldehyde, 6-methylpyridyl-2-aldehyde
70737
6-methylpyridine-2-carbaldehyde
CC1=CC=CC(=N1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 31°C to 33°C
Color Yellow to Brown
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH(C5H3N)COH
Refractive Index 1.527
Quantity 1 g
Solubility Information Solubility in water: 750g/L water (20°C). Other solubilities: soluble in ether and acetone
Formula Weight 121.14
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 6-Methyl-2-pyridinecarboxaldehyde, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methyl-2-pyridinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'