missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-methyl-2,3-dihydropyridazin-3-one hydrate, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
Assay Percent Range | 90% |
Quantity | 1 g |
IUPAC Name | 6-methyl-2,3-dihydropyridazin-3-one |
Formula Weight | 128.14 |
Percent Purity | 90% |
Chemical Name or Material | 6-Methyl-2, 3-dihydropyridazin-3-one hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'