missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Methyl-2-(2-thienyl)benzimidazole, 97%

Brand:  Alfa Aesar™ H58782.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15484988

  • £725.99 / 5g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

6-Methyl-2-(2-thienyl)benzimidazole
214.28
MFCD14618840

Specifications

6-Methyl-2-(2-thienyl)benzimidazole
5g
97%
MFCD14618840
Yellow to pale brown
Solid
214.28
Videos
SDS