missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxytryptamine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
250 mg
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 143.0°C to 146.0°C |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 50 mg |
Synonym | 6-methoxytryptamine, 2-6-methoxy-1h-indol-3-yl ethanamine, 3-2-aminoethyl-6-methoxyindole, 2-6-methoxy-1h-indol-3-yl-ethylamine, 1h-indole-3-ethanamine, 6-methoxy, 2-2-aminoethyl-5-methoxyindole, 1h-indole-3-ethanamine,6-methoxy, 2-6-methoxy-1h-indol-3-yl ethan-1-amine, 2-6-methoxyindol-3-yl ethylamine, 6-methoxy tryptamine |
Solubility Information | Solubility in water: moderately soluble. Other solubilities: soluble in ethanol |
Formula Weight | 190.24 |
Percent Purity | 99% |
Show More |
Safety and Handling
Product Identifier
- 6-Methoxytryptamine
- Danger
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'