missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Methoxyquinoxaline, 98%

Brand:  Alfa Aesar™ H63396.03

Code : NEW

Additional Details : CAS Number : 6639-82-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450069

  • £49.00 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

6-Methoxyquinoxaline
6639-82-3
160.17
MFCD00666601

Specifications

6-Methoxyquinoxaline
Solid
160.17
1g
6639-82-3
98%
MFCD00666601
Videos
SDS