missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1-tetralone, 99%, Thermo Scientific Chemicals

Catalog Number 10793701
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1078-19-9
C11H12O2
176.21
MFCD00001695
MNALUTYMBUBKNX-UHFFFAOYSA-N
6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone
14112
6-methoxy-3,4-dihydro-2H-naphthalen-1-one
COC1=CC2=C(C=C1)C(=O)CCC2
This item is not returnable. View return policy
1078-19-9
C11H12O2
176.21
MFCD00001695
MNALUTYMBUBKNX-UHFFFAOYSA-N
6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone
14112
6-methoxy-3,4-dihydro-2H-naphthalen-1-one
COC1=CC2=C(C=C1)C(=O)CCC2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 77.0°C to 79.0°C
Color Brown to Yellow
Boiling Point 171.0°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 09, II, 889
Merck Index 15, 6074
Formula Weight 176.21
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 6-Methoxy-1-tetralone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'