missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Maleimidohexanoic acid, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. Further, it is used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. In addition to this, it acts as a probe for thiol groups in membrane proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Yellow |
Quantity | 100 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water,methanol,ethanol,dimethyl sulfoxide. Insoluble in ether. |
Formula Weight | 211.22 |
Physical Form | Powder |
Chemical Name or Material | 6-Maleimidohexanoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'