missing translation for 'onlineSavingsMsg'
Learn More

6-Maleimidohexanoic acid, Thermo Scientific Chemicals

Catalog Number 15472677
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
55750-53-3
C10H13NO4
211.217
MFCD00043140
WOJKKJKETHYEAC-UHFFFAOYSA-N
6-maleimidocaproic acid, 6-maleimidohexanoic acid, 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoic acid, 6-maleimidocaproicacid, 1h-pyrrole-1-hexanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 5-cooh, n-maleoyl-6-aminocaproic acid, n-maleoyl-6-aminohexanoic acid, 1h-pyrrole-1-hexanoicacid, 2,5-dihydro-2,5-dioxo, 6-2,5-dioxopyrrol-1-yl hexanoic acid
573683
6-(2,5-dioxopyrrol-1-yl)hexanoic acid
C1=CC(=O)N(C1=O)CCCCCC(=O)O
This item is not returnable. View return policy
55750-53-3
C10H13NO4
211.217
MFCD00043140
WOJKKJKETHYEAC-UHFFFAOYSA-N
6-maleimidocaproic acid, 6-maleimidohexanoic acid, 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoic acid, 6-maleimidocaproicacid, 1h-pyrrole-1-hexanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 5-cooh, n-maleoyl-6-aminocaproic acid, n-maleoyl-6-aminohexanoic acid, 1h-pyrrole-1-hexanoicacid, 2,5-dihydro-2,5-dioxo, 6-2,5-dioxopyrrol-1-yl hexanoic acid
573683
6-(2,5-dioxopyrrol-1-yl)hexanoic acid
C1=CC(=O)N(C1=O)CCCCCC(=O)O

6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. Further, it is used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. In addition to this, it acts as a probe for thiol groups in membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,methanol,ethanol,dimethyl sulfoxide. Insoluble in ether.
Formula Weight 211.22
Physical Form Powder
Chemical Name or Material 6-Maleimidohexanoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'