missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-1H-indazole, 97%, Thermo Scientific™

Catalog Number p-4228746
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
261953-36-0
C7H5IN2
244.03
MFCD04114695
RSGAXJZKQDNFEP-UHFFFAOYSA-N
6-iodoindazole, 1h-indazole, 6-iodo, 6-iodo 1h indazole, 6-iodo-indazole, pubchem18081, indazole, 6-iodo
12991241
6-iodo-1H-indazole
C1=CC2=C(C=C1I)NN=C2
This item is not returnable. View return policy
261953-36-0
C7H5IN2
244.03
MFCD04114695
RSGAXJZKQDNFEP-UHFFFAOYSA-N
6-iodoindazole, 1h-indazole, 6-iodo, 6-iodo 1h indazole, 6-iodo-indazole, pubchem18081, indazole, 6-iodo
12991241
6-iodo-1H-indazole
C1=CC2=C(C=C1I)NN=C2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 208°C to 211°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 244.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Iodo-1H-indazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'