missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxypyridine-2-carboxylic acid, 95%, ACROS Organics™

Brand:  Acros Organics 434820250

Code : 29

Additional Details : CAS Number : 19621-92-2 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10862241

  • £128.60 / 25g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

6-Hydroxypyridine-2-carboxylic acid
94.0
≥94%
C6H5NO3
MFCD00192220
19621-92-2
100.0
95%
139.11
6-Hydroxypicolinic acid

Specifications

6-Hydroxypyridine-2-carboxylic acid
25g
94.0
Authentic
Powder
95%
139.11
6-Hydroxypicolinic acid
Yellow to brown
19621-92-2
100.0
Glass bottle
≥94%
C6H5NO3
MFCD00192220
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Hydroxypyridine-2-carboxylic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS