missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Hydroxynaphthalene-2-boronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62631.03

Code : NEW

Additional Details : CAS Number : 269410-21-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15437656

  • £172.80 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

6-Hydroxynaphthalene-2-boronic acid pinacol ester
270.13
6-Hydroxy-2-naphthaleneboronic acid pinacol ester
269410-21-1
MFCD20486767

Specifications

6-Hydroxynaphthalene-2-boronic acid pinacol ester
1g
Solid
270.13
6-Hydroxy-2-naphthaleneboronic acid pinacol ester
White
269410-21-1
96%
MFCD20486767
Videos
SDS