missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Hydroxymethyl)benzothiazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 103°C to 106°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 165.22 |
Percent Purity | ≥96.0% |
Physical Form | Crystalline Powder or Needles |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'