missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Hydroxyindole, 98%

Brand:  Alfa Aesar™ H55951.03

Code : NEW

Additional Details : CAS Number : 2380-86-1 Weight : 0.00100kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15402358

  • £129.00 / 1g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2380-86-1
133.15
XAWPKHNOFIWWNZ-UHFFFAOYSA-N
524508
C1=CC(=CC2=C1C=CN2)O
C8H7NO
MFCD00152101
6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol
1H-indol-6-ol

Specifications

6-Hydroxyindole
1g
C8H7NO
UN3077
XAWPKHNOFIWWNZ-UHFFFAOYSA-N
1H-indol-6-ol
524508
98%
125°C to 128°C
2380-86-1
MFCD00152101
6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol
C1=CC(=CC2=C1C=CN2)O
133.15
133.15
Videos
SDS Documents
Certificates