missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-3-nitro-2-picoline, 98%, Thermo Scientific™

Catalog Number p-3778831
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
28489-45-4
MFCD00092010
AJDDHNXZBGZBJN-UHFFFAOYSA-N
6-methyl-5-nitropyridin-2-ol, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 2-hydroxy-5-nitro-6-picoline, 6-hydroxy-2-methyl-3-nitropyridine, 6-methyl-5-nitropyridin-2 1h-one, 2 1h-pyridinone, 6-methyl-5-nitro, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, pubchem10387
543053
6-methyl-5-nitro-1H-pyridin-2-one
CC1=C(C=CC(=O)N1)[N+](=O)[O-]
This item is not returnable. View return policy
28489-45-4
MFCD00092010
AJDDHNXZBGZBJN-UHFFFAOYSA-N
6-methyl-5-nitropyridin-2-ol, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 2-hydroxy-5-nitro-6-picoline, 6-hydroxy-2-methyl-3-nitropyridine, 6-methyl-5-nitropyridin-2 1h-one, 2 1h-pyridinone, 6-methyl-5-nitro, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, pubchem10387
543053
6-methyl-5-nitro-1H-pyridin-2-one
CC1=C(C=CC(=O)N1)[N+](=O)[O-]
CAS Min % 97.5
CAS Max % 100.0
Melting Point 230°C to 232°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H6N2O3
Quantity 1 g
Molecular Weight (g/mol) 154
Formula Weight 154
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-3-nitro-2-picoline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 6-Hydroxy-3-nitro-2-picoline
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'