missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoropyridine-3-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog Number p-4813264
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
677728-92-6
C6H4FNO
125.1
PZPNGWWKCSJKOS-UHFFFAOYSA-N
2-fluoropyridine-5-carbaldehyde, 6-fluoronicotinaldehyde, 2-fluoro-5-formylpyridine, 2-fluoropyridine-5-carboxaldehyde, 6-fluoro-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 6-fluoro, 6-fluoropyridine-3-carboxaldehyde, pubchem5150, zlchem 1162, acmc-1b2mv
16414246
6-fluoropyridine-3-carbaldehyde
C1=CC(=NC=C1C=O)F
This item is not returnable. View return policy
677728-92-6
C6H4FNO
125.1
PZPNGWWKCSJKOS-UHFFFAOYSA-N
2-fluoropyridine-5-carbaldehyde, 6-fluoronicotinaldehyde, 2-fluoro-5-formylpyridine, 2-fluoropyridine-5-carboxaldehyde, 6-fluoro-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 6-fluoro, 6-fluoropyridine-3-carboxaldehyde, pubchem5150, zlchem 1162, acmc-1b2mv
16414246
6-fluoropyridine-3-carbaldehyde
C1=CC(=NC=C1C=O)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 125.1
Percent Purity 95%
Chemical Name or Material 6-Fluoropyridine-3-carboxaldehyde
compliance-icons
Product Identifier
  • 6-Fluoropyridine-3-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'