missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoropyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4258860
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
402-69-7
C6H4FNO2
141.10
MFCD02181193
DIEMCUFYSOEIDU-UHFFFAOYSA-N
6-fluoropicolinic acid, 2-fluoropyridine-6-carboxylic acid, 6-fluoro-2-pyridinecarboxylic acid, 2-fluoro-6-pyridinecarboxylic acid, 2-fluoro-6-carboxypyridine, 6-fluoro-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-fluoro, 6-fluor-2-pyridincarbons, pubchem13570, 6-fluoro picolinic acid
226027
6-fluoropyridine-2-carboxylic acid
OC(=O)C1=CC=CC(F)=N1
This item is not returnable. View return policy
402-69-7
C6H4FNO2
141.10
MFCD02181193
DIEMCUFYSOEIDU-UHFFFAOYSA-N
6-fluoropicolinic acid, 2-fluoropyridine-6-carboxylic acid, 6-fluoro-2-pyridinecarboxylic acid, 2-fluoro-6-pyridinecarboxylic acid, 2-fluoro-6-carboxypyridine, 6-fluoro-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-fluoro, 6-fluor-2-pyridincarbons, pubchem13570, 6-fluoro picolinic acid
226027
6-fluoropyridine-2-carboxylic acid
OC(=O)C1=CC=CC(F)=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 138.4°C to 141.1°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 141.1
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 6-Fluoropyridine-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Fluoropyridine-2-carboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'