missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Fluoro-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole, 97%

Brand:  Alfa Aesar™ H63764.03

Code : NEW

Additional Details : CAS Number : 200714-09-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458506

  • £59.78 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

6-Fluoro-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
200714-09-6
230.28
MFCD28126777

Specifications

6-Fluoro-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
200714-09-6
97%
MFCD28126777
1g
Solid
230.28
Videos
SDS