missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Fluoro-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole, 97%

Brand:  Alfa Aesar™ H63764.06

Code : NEW

Additional Details : CAS Number : 200714-09-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15468506

  • £298.92 / 5g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

6-Fluoro-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
200714-09-6
230.28
MFCD28126777

Specifications

6-Fluoro-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
200714-09-6
97%
MFCD28126777
5g
Solid
230.28
Videos
SDS