missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2,3-dihydro-4H-thiochromen-4-one, ≥95%, Thermo Scientific™

Catalog Number p-2702795
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
21243-18-5
C9H7FOS
182.212
MFCD00078488
NBBGHADNMPMHST-UHFFFAOYSA-N
6-fluorothio-4-chromanone, 6-fluoro-2,3-dihydro-4h-thiochromen-4-one, 6-fluorothiochroman-4-one, 6-fluoro-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-fluoro-3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,6-fluoro-2,3-dihydro, 6-fluoro-3,4-dihydro-2h-1-benzothiin-4-one, 6-fluoro-2,3-dihydro-1-benzothiopyran-4-one, maybridge1_007680, 6-fluoro-4-oxothiochroman
2737521
6-fluoro-2,3-dihydrothiochromen-4-one
C1CSC2=C(C1=O)C=C(C=C2)F
This item is not returnable. View return policy
21243-18-5
C9H7FOS
182.212
MFCD00078488
NBBGHADNMPMHST-UHFFFAOYSA-N
6-fluorothio-4-chromanone, 6-fluoro-2,3-dihydro-4h-thiochromen-4-one, 6-fluorothiochroman-4-one, 6-fluoro-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-fluoro-3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,6-fluoro-2,3-dihydro, 6-fluoro-3,4-dihydro-2h-1-benzothiin-4-one, 6-fluoro-2,3-dihydro-1-benzothiopyran-4-one, maybridge1_007680, 6-fluoro-4-oxothiochroman
2737521
6-fluoro-2,3-dihydrothiochromen-4-one
C1CSC2=C(C1=O)C=C(C=C2)F
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 182.22
Percent Purity 95%
Chemical Name or Material 6-Fluoro-2, 3-dihydro-4H-thiochromen-4-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'