missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-1H-indazole-3-carboxylic acid, 97%, Acros Organics

Brand:  Acros Organics 450390050

UNSPSC : 12352100

Code : NEW

Additional Details : CAS Number : 129295-30-3 Weight : 0.00500kg


Quantity 5g

 View more versions of this product

Product Code. 15234824

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

6-Fluoro-1H-indazole-3-carboxylic acid
97%
C8H5FN2O2
180.14

Specifications

6-Fluoro-1H-indazole-3-carboxylic acid
129295-30-3
C8H5FN2O2
5g
97%
180.14
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Fluoro-1H-indazole-3-carboxylic acid, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.