missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Cyano-3-(Dimethylaminomethyl)indole, 95%

Brand:  Alfa Aesar™ H63612.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15410399

  • £95.61 / 1g


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

6-Cyano-3-(Dimethylaminomethyl)indole
MFCD11112020
14198963
199.25
UN3439
6-Cyanogramine; 3-(Dimethylaminomethyl)indole-6-carbonitrile

Specifications

6-Cyano-3-(Dimethylaminomethyl)indole
Solid
199.25
UN3439
6-Cyanogramine; 3-(Dimethylaminomethyl)indole-6-carbonitrile
1g
95%
MFCD11112020
14198963
Off-white to brown
Videos
SDS