missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Cyano-3-(1,2,3,6-tetrahydro-4-pyridyl)indole, 97%

Brand:  Alfa Aesar™ H63388.MD

Code : NEW

Additional Details : CAS Number : 918525-20-9 Weight : 0.00025kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15468286

  • £35.83 / 250mg


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

6-Cyano-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
223.27
UN3439
918525-20-9
MFCD28098703
3-(1,2,3,6-Tetrahydro-4-pyridyl)indole-6-carbonitrile

Specifications

6-Cyano-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
250mg
Solid
223.27
UN3439
Off-white to yellow
918525-20-9
97%
MFCD28098703
3-(1,2,3,6-Tetrahydro-4-pyridyl)indole-6-carbonitrile
Videos
SDS