Learn More
Alfa Aesar™ 6-Cyano-2-(3-pyridyl)benzimidazole, 97%
Brand: Alfa Aesar™ H58929.03
Code : NEW
Additional Details : Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
6-Cyano-2-(3-pyridyl)benzimidazole | |
220.23 | |
MFCD22581247 | |
UN3439 |
Specifications
6-Cyano-2-(3-pyridyl)benzimidazole | |
234° to 240°C | |
Powder | |
220.23 | |
UN3439 |
Pale brown to brown | |
1g | |
97% | |
MFCD22581247 |