missing translation for 'onlineSavingsMsg'
Learn More

6-Cyano-1-methylindole, 97%, Thermo Scientific™

Catalog Number 15470779
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
20996-87-6
C10H8N2
156.188
MFCD08690254
FOGAAHLMYPQCJG-UHFFFAOYSA-N
1-methyl-1h-indole-6-carbonitrile, 1h-indole-6-carbonitrile, 1-methyl, 6-cyano-1-methylindole, acmc-1cirg, 1-methyl-6-cyanoindole, 6-cyano-1-methyl-1h-indole, 1h-indole-6-carbonitrile, 1-methyl-9ci
15384493
1-methylindole-6-carbonitrile
CN1C=CC2=C1C=C(C=C2)C#N
This item is not returnable. View return policy
20996-87-6
C10H8N2
156.188
MFCD08690254
FOGAAHLMYPQCJG-UHFFFAOYSA-N
1-methyl-1h-indole-6-carbonitrile, 1h-indole-6-carbonitrile, 1-methyl, 6-cyano-1-methylindole, acmc-1cirg, 1-methyl-6-cyanoindole, 6-cyano-1-methyl-1h-indole, 1h-indole-6-carbonitrile, 1-methyl-9ci
15384493
1-methylindole-6-carbonitrile
CN1C=CC2=C1C=C(C=C2)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Quantity 250 mg
UN Number UN3439
Beilstein 472731
Formula Weight 156.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Cyano-1-methylindole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'