missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Cyano-1-methylindole, 97%

Brand:  Alfa Aesar™ H63853.06

Code : NEW

Additional Details : CAS Number : 20996-87-6 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15460779

  • £232.40 / 5g


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

6-Cyano-1-methylindole
156.18
UN3439
1-Methylindole-6-carbonitrile
20996-87-6
MFCD08690254
472731

Specifications

6-Cyano-1-methylindole
5g
Solid
156.18
UN3439
1-Methylindole-6-carbonitrile
Off-white to yellow
20996-87-6
97%
MFCD08690254
472731
Videos
SDS