missing translation for 'onlineSavingsMsg'
Learn More

6-chloropyridine-2-carboxylic acid, 97%, ACROS Organics™

Brand:  Acros Organics 436660050

Code : 29

Additional Details : CAS Number : 4684-94-0 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10493310

  • £88.40 / 5g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

6-Chloropyridine-2-carboxylic acid
96.0
≥96.0%
C6H4ClNO2
4684-94-0
100.0
97%
157.56

Specifications

6-Chloropyridine-2-carboxylic acid
160° to 164°C
4684-94-0
100.0
Glass bottle
≥96.0%
C6H4ClNO2
White to cream to tan
5g
96.0
Authentic
Crystalline Powder
97%
157.56
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Chloropyridine-2-carboxylic acid, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS