missing translation for 'onlineSavingsMsg'
Learn More

6-chloropyridazine-3-carboxylic acid, 97%, ACROS Organics™

Brand:  Acros Organics 429530050

Code : 29

Additional Details : CAS Number : 5096-73-1 Weight : 0.06000kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10116297

  • £399.20 / 5g


Estimated Shipment
21-06-2018
Add to basket

Specifications

Chemical Identifiers

6-chloropyridazine-3-carboxylic acid
96.0
97%
C5H3ClN2O2
MFCD00160464
5096-73-1
100.0
97%
158.54

Specifications

6-chloropyridazine-3-carboxylic acid
145.0° to 148.0°C
5096-73-1
100.0
Glass bottle
97%
C5H3ClN2O2
MFCD00160464
White to cream
5g
96.0
Authentic
Crystalline Powder
97%
158.54
Videos
SDS