missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine, 99%, Thermo Scientific Chemicals

Catalog Number p-7081291
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
87-42-3
C5H3ClN4
154.557
MFCD00075825
ZKBQDFAWXLTYKS-UHFFFAOYSA-N
6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
5359277
6-chloro-7H-purine
C1=NC2=C(N1)C(=NC=N2)Cl
This item is not returnable. View return policy
87-42-3
C5H3ClN4
154.557
MFCD00075825
ZKBQDFAWXLTYKS-UHFFFAOYSA-N
6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
5359277
6-chloro-7H-purine
C1=NC2=C(N1)C(=NC=N2)Cl

6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.

Solubility
Soluble in hot water and dimethyl formamide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point >300°C (decomposition)
Quantity 1 g
Beilstein 5774
Merck Index 14,2161
Solubility Information Soluble in hot water and dimethyl formamide.
Formula Weight 154.56
Percent Purity 99%
Chemical Name or Material 6-Chloropurine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'