missing translation for 'onlineSavingsMsg'
Learn More

6-chloronicotinamide, 98%, Thermo Scientific™

Catalog Number p-4323392
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6271-78-9
C6H5ClN2O
156.57
MFCD00006242
ZIJAZUBWHAZHPL-UHFFFAOYSA-N
6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t
80456
6-chloropyridine-3-carboxamide
NC(=O)C1=CC=C(Cl)N=C1
This item is not returnable. View return policy
6271-78-9
C6H5ClN2O
156.57
MFCD00006242
ZIJAZUBWHAZHPL-UHFFFAOYSA-N
6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t
80456
6-chloropyridine-3-carboxamide
NC(=O)C1=CC=C(Cl)N=C1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 204.4°C to 207.9°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula Cl(C5H3N)CONH2
Quantity 1 g
Beilstein 22,44
Formula Weight 156.57
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 6-Chloronicotinamide
Show More Show Less
compliance-icons
Product Identifier
  • 6-Chloronicotinamide, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'