missing translation for 'onlineSavingsMsg'
Learn More

6-(chloromethyl)benzothiazole, 97%, Thermo Scientific™

Catalog Number p-4258899
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
149440-49-3
C8H6ClNS
183.66
WVJSICZOZHJCLJ-UHFFFAOYSA-N
6-chloromethyl benzo d thiazole, 6-chloromethyl benzothiazole, benzothiazole, 6-chloromethyl, 6-chloromethyl-1,3-benzothiazole, acmc-20n5pd, 6-chloromethylbenzothiazole
22905771
6-(chloromethyl)-1,3-benzothiazole
C1=CC2=C(C=C1CCl)SC=N2
This item is not returnable. View return policy
149440-49-3
C8H6ClNS
183.66
WVJSICZOZHJCLJ-UHFFFAOYSA-N
6-chloromethyl benzo d thiazole, 6-chloromethyl benzothiazole, benzothiazole, 6-chloromethyl, 6-chloromethyl-1,3-benzothiazole, acmc-20n5pd, 6-chloromethylbenzothiazole
22905771
6-(chloromethyl)-1,3-benzothiazole
C1=CC2=C(C=C1CCl)SC=N2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 52°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 183.66
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-(Chloromethyl)benzothiazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 6-(Chloromethyl)benzothiazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'