Learn More
6-Chloromelatonin, Tocris Bioscience™
Melatonin agonist
Brand:  Tocris (Bio-Techne) 044310
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487653
  • £92.00 / 10mg
Estimated Shipment
26-06-2018
Add to basket
Description
Description
Potent melatonin agonist (pKi values are 9.10 and 9.77 for human recombinant MT1 and MT2 receptors respectively). Displays higher affinity for binding to hamster brain membrane and chicken retina than melatonin.
Specifications
Chemical Identifiers
6-Chloromelatonin
C13H15ClN2O2
266.73
N-[2-(6-Chloro-5-methoxyindol-3-yl)ethyl]acetamide
Specifications
6-Chloromelatonin
63762-74-3
C13H15ClN2O2
N-[2-(6-Chloro-5-methoxyindol-3-yl)ethyl]acetamide
10mg
>99%
266.73
Videos