missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Chloroindole-3-carboxaldehyde, 98%

Brand:  Alfa Aesar™ H61911.14

Code : NEW

Additional Details : CAS Number : 703-82-2 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15418508

  • £139.90 / 25g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Description

Description

Applications
6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

6-Chloroindole-3-carboxaldehyde
703-82-2
179.6
MFCD06657152

Specifications

6-Chloroindole-3-carboxaldehyde
25g
Powder
179.6
Pale brown
703-82-2
98%
MFCD06657152
Videos
SDS