missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-4510544
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
150020-64-7
C5H2Cl2N2O2S2
257.103
APRATEMYDZDYJD-UHFFFAOYSA-N
2799717
6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
C1=CSC2=NC(=C(N21)S(=O)(=O)Cl)Cl
This item is not returnable. View return policy
150020-64-7
C5H2Cl2N2O2S2
257.103
APRATEMYDZDYJD-UHFFFAOYSA-N
2799717
6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
C1=CSC2=NC(=C(N21)S(=O)(=O)Cl)Cl
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro-imidazo 2,1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro, acmc-1c5af, 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonylchloride, 6-chloro-5-imidazo 2,1-b thiazolesulfonyl chloride, 6-chloro-imidazo 2, 1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride,6-chloro
Formula Weight 257.12
Percent Purity 97%
Chemical Name or Material 6-Chloroimidazo[2, 1-b][1, 3]thiazole-5-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'