missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Chloro-9-(2-tetrahydropyranyl)purine, 99%

Brand:  Alfa Aesar™ H63779.30

Code : NEW

Additional Details : CAS Number : 7306-68-5 Weight : 0.25000kg

Quantity 250g

 View more versions of this product

Product Code. 15450659

  • £117.77 / 250g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
6-Chloro-9-(2-tetrahydropyranyl)purine is used to prepare 6-anilino-9-(2-tetrahydropyranyl)purine by reacting with bromobenzene in the presence of potassium amide as a reagent. Further, it serves as an intermediate in organic synthesis and pharmaceuticals.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

6-Chloro-9-(2-tetrahydropyranyl)purine
7306-68-5
238.68
MFCD00022821

Specifications

6-Chloro-9-(2-tetrahydropyranyl)purine
White to pale yellow
250g
1.7410
99%
MFCD00022821
1.604g/mL
149° to 151°C
7306-68-5
Solid
238.68
Videos
SDS