missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Chloro-5-fluorobenzimidazole, 95%

Brand:  Alfa Aesar™ A10904.03

Code : NEW

Additional Details : CAS Number : 175135-04-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11464273

  • £47.90 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
6-Chloro-5-fluorobenzimidazole is used as a pharamceutical intermediate, agrochemicals. As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment; subsequently several of the top pharmaceutical drugs contain fluorine.

Solubility
Insoluble in both water and organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

6-Chloro-5-fluorobenzimidazole
C7H4ClFN2
MFCD00156132
175135-04-3
170.56

Specifications

6-Chloro-5-fluorobenzimidazole
1g
95%
170.56
148° to 150°C
175135-04-3
C7H4ClFN2
MFCD00156132
Videos
SDS