missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-2-tetralone, 97%, Thermo Scientific™

Catalog Number 10622455
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17556-18-2
C10H9ClO
180.63
MFCD03788443
QQSYTUUAEZUAKL-UHFFFAOYSA-N
6-chloro-2-tetralone, 6-chloro-3,4-dihydronaphthalen-2 1h-one, 2 1h-naphthalenone, 6-chloro-3,4-dihydro, 6-chloro-3,4-dihydro-2 1h-naphthalenone, 2 1h-naphthalenone,6-chloro-3,4-dihydro, pubchem17950, 6-chloro-2-tetrarlone, acmc-20a1a6, 6-chloro-beta-tetralone, 6-chloro-3.4-dihydro-2 1h naphthalenone
4456139
ClC1=CC2=C(CC(=O)CC2)C=C1
This item is not returnable. View return policy
17556-18-2
C10H9ClO
180.63
MFCD03788443
QQSYTUUAEZUAKL-UHFFFAOYSA-N
6-chloro-2-tetralone, 6-chloro-3,4-dihydronaphthalen-2 1h-one, 2 1h-naphthalenone, 6-chloro-3,4-dihydro, 6-chloro-3,4-dihydro-2 1h-naphthalenone, 2 1h-naphthalenone,6-chloro-3,4-dihydro, pubchem17950, 6-chloro-2-tetrarlone, acmc-20a1a6, 6-chloro-beta-tetralone, 6-chloro-3.4-dihydro-2 1h naphthalenone
4456139
ClC1=CC2=C(CC(=O)CC2)C=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 61.0°C to 65.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
IUPAC Name 6-chloro-1,2,3,4-tetrahydronaphthalen-2-one
Formula Weight 180.63
Percent Purity 97%
Chemical Name or Material 6-chloro-2-tetralone
compliance-icons
Product Identifier
  • 6-Chloro-2-tetralone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'