missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-2-picoline, 98%, Thermo Scientific Chemicals

Catalog Number 10771414
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18368-63-3
C6H6ClN
127.57
MFCD00006245
GXZDYRYYNXYPMQ-UHFFFAOYSA-N
6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
87601
2-chloro-6-methylpyridine
CC1=CC=CC(Cl)=N1
This item is not returnable. View return policy
18368-63-3
C6H6ClN
127.57
MFCD00006245
GXZDYRYYNXYPMQ-UHFFFAOYSA-N
6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
87601
2-chloro-6-methylpyridine
CC1=CC=CC(Cl)=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 64°C to 68°C (10.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(C5H3N)Cl
Refractive Index 1.5260 to 1.528
Quantity 50 g
Beilstein 20, I, 84
Specific Gravity 1.16
Formula Weight 127.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 6-chloro-2-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 6-Chloro-2-picoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'