missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromopyridine-2-carbaldehyde, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 186.01 |
Percent Purity | 97% |
Chemical Name or Material | 6-Bromopyridine-2-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'