missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyridine-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7029748
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
651358-83-7
C11H15BBrNO2
283.96
MFCD06796177
OAVRLCKBKDMGGW-UHFFFAOYSA-N
6-bromopyridine-2-boronic acid pinacol ester, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester
12177444
2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)Br
This item is not returnable. View return policy
651358-83-7
C11H15BBrNO2
283.96
MFCD06796177
OAVRLCKBKDMGGW-UHFFFAOYSA-N
6-bromopyridine-2-boronic acid pinacol ester, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester
12177444
2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 125°C to 127°C
Quantity 250 mg
Formula Weight 283.96
Percent Purity 95%
Chemical Name or Material 6-Bromopyridine-2-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'